4-Fluorphenylhydraziniumchlorid
Lieferant: Thermo Scientific
Synonyme:
4-Fluorphenylhydrazin Hydrochlorid, 1-(4-Fluorphenyl)hydrazin Hydrochlorid, 4-Fluorphenylhydrazinhydrochlorid
BTB08671.1
BTB08671.50
MAYBBTB08671.1EA
0
EUR
MAYBBTB08671.1
MAYBBTB08671.50
4-Fluorphenylhydraziniumchlorid
4-Fluorphenylhydraziniumchlorid
Formel:
C₆H₈ClFN₂ Molecular Weight: 162.59 g/mol |
MDL:
MFCD00012942 CAS-Nummer: 823-85-8 EINECS: 212-521-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...