4-Bromphenylhydraziniumchlorid

Lieferant: CHEMPUR

BTB08672.1 BTB08672.10 BTB08672.50
CHMPBTB08672.1EA 0 EUR
CHMPBTB08672.1 CHMPBTB08672.10 CHMPBTB08672.50
4-Bromphenylhydraziniumchlorid
4-Bromphenylhydraziniumchlorid
Formel: C₆H₈BrClN₂
MDL: MFCD00012941
CAS-Nummer: 622-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR