Methyl-2-brom-5-methoxybenzoat
Lieferant: Thermo Scientific
Synonyme:
2-Brom-5-methoxybenzoesäuremethylester
CD05257.50
CD05257.1
MAYBCD05257.50EA
0
EUR
MAYBCD05257.50
MAYBCD05257.1
Methyl-2-brom-5-methoxybenzoat
Methyl-2-brom-5-methoxybenzoat
Formel:
C₉H₉BrO₃ Molecular Weight: 245.07 g/mol |
MDL:
MFCD00051594 CAS-Nummer: 35450-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...