2-(Trifluormethyl)phenylhydrazin

Lieferant: Thermo Scientific

Synonyme: 1-(2-(Trifluormethyl)phenyl)hydrazin, 2-Hydrazinobenzotrifluorid

RF01370DA RF01370EA
MAYBRF01370DAEA 0 EUR
MAYBRF01370DA MAYBRF01370EA
2-(Trifluormethyl)phenylhydrazin
2-(Trifluormethyl)phenylhydrazin
Formel: C₇H₇F₃N₂
Molecular Weight: 176,14 g/mol
MDL: MFCD00052687
CAS-Nummer: 365-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR