Propargylamin Hydrochlorid

Lieferant: Thermo Scientific

Synonyme: 2-Propinylammoniumchlorid, Propargylammoniumchlorid, Monopropargylamin Hydrochlorid, Prop-2-ynylamin Hydrochlorid, 3-Aminopropin Hydrochlorid

RH00060.1 RH00060.10 RH00060.50
MAYBRH00060.1EA 0 EUR
MAYBRH00060.1 MAYBRH00060.10 MAYBRH00060.50
Propargylamin Hydrochlorid
Propargylamin Hydrochlorid
Formel: C₃H₆ClN
Schmelzpunkt: 178…179 °C
MDL: MFCD00012907
CAS-Nummer: 15430-52-1
EINECS: 239-440-5
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR