2-Butyl-1,3-propandiol

Lieferant: CHEMPUR

Synonyme: 2-Butylpropane-1,3-diol

RJC03456.250 RJC03456.1
CHMPRJC03456.250EA 0 EUR
CHMPRJC03456.250 MAYBRJC03456.1
2-Butyl-1,3-propandiol
2-Butyl-1,3-propanediol
Formel: C₇H₁₆O₂
Molecular Weight: 132,2 g/mol
MDL: MFCD00799395
CAS-Nummer: 2612-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR