Methyl-2-amino-4,5-dimethoxybenzoat

Lieferant: Thermo Scientific

Synonyme: Methyl-4,5-dimethoxyanthranilat, 2-Amino-4,5-dimethoxybenzoesäuremethylester

SB01989DA SB01989.10 SB01989.50
MAYBSB01989DAEA 0 EUR
MAYBSB01989DA MAYBSB01989.10 MAYBSB01989.50
Methyl-2-amino-4,5-dimethoxybenzoat
Methyl-2-amino-4,5-dimethoxybenzoat
Formel: C₁₀H₁₃NO₄
Molecular Weight: 211.22 g/mol
MDL: MFCD00014904
CAS-Nummer: 26759-46-6
EINECS: 247-967-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR