3-Phenoxy-2,4-pentandion
Lieferant: Thermo Scientific
SPB05304.1
SPB05304.10
MAYBSPB05304.1EA
0
EUR
MAYBSPB05304.1
MAYBSPB05304.10
3-Phenoxy-2,4-pentandion
3-Phenoxy-2,4-pentandion
Formel:
C₁₁H₁₂O₃ |
CAS-Nummer:
31614-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...