2-(Trifluormethyl)phenylhydraziniumchlorid

Lieferant: Thermo Scientific

Synonyme: 2-(Trifluormethyl)phenylhydrazin Hydrochlorid, 2-(Trifluormethyl)phenylhydrazinhydrochlorid, 2-Hydrazinobenzotrifluorid Hydrochlorid, 1-(2-(Trifluormethyl)phenyl)hydrazin Hydrochlorid

SPB06067.1 SPB06067EA
MAYBSPB06067.1EA 0 EUR
MAYBSPB06067.1 MAYBSPB06067EA
2-(Trifluormethyl)phenylhydraziniumchlorid
2-(Trifluormethyl)phenylhydraziniumchlorid
Formel: C₇H₈ClF₃N₂
Molecular Weight: 212,6 g/mol
MDL: MFCD00102619
CAS-Nummer: 3107-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR