4-(Trifluormethyl)phenylhydraziniumchlorid

Lieferant: Thermo Scientific

Synonyme: 1-(4-(Trifluormethyl)phenyl)hydrazin Hydrochlorid, 4-(Trifluormethyl)phenylhydrazin Hydrochlorid, 4-Hydrazinobenzotrifluorid Hydrochlorid, 4-(Trifluormethyl)phenylhydrazinhydrochlorid

SPB06880DA SPB06880EA
MAYBSPB06880DAEA 0 EUR
MAYBSPB06880DA MAYBSPB06880EA
4-(Trifluormethyl)phenylhydraziniumchlorid
4-(Trifluormethyl)phenylhydraziniumchlorid
Formel: C₇H₈ClF₃N₂
Molecular Weight: 212,6 g/mol
MDL: MFCD00204233
CAS-Nummer: 2923-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR