Methyl-3-amino-4-methoxybenzoat
Lieferant: Thermo Scientific
Synonyme:
3-Amino-4-methoxybenzoesäuremethylester
TL00302DA
TL00302.10
TL00302.50
MAYBTL00302DAEA
0
EUR
MAYBTL00302DA
MAYBTL00302.10
MAYBTL00302.50
Methyl-3-amino-4-methoxybenzoat
Methyl-3-amino-4-methoxybenzoat
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol |
MDL:
MFCD00017196 CAS-Nummer: 24812-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...