Methyl-5-chlor-2-methoxybenzoat
Lieferant: Thermo Scientific
Synonyme:
5-Chlor-2-methoxybenzoesäuremethylester
TL00590.1
TL00590.10
TL00590.50
MAYBTL00590.1EA
0
EUR
MAYBTL00590.1
MAYBTL00590.10
MAYBTL00590.50
Methyl-5-chlor-2-methoxybenzoat
Methyl-5-chlor-2-methoxybenzoat
Formel:
C₉H₉ClO₃ Molecular Weight: 200,62 g/mol |
MDL:
MFCD00017511 CAS-Nummer: 33924-48-0 EINECS: 251-743-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...