Methyl-4-methoxy-3-nitrobenzoat
Lieferant: CHEMPUR
Synonyme:
4-Methoxy-3-nitrobenzoesäuremethylester
TL00923.250
TL00923.1
CHMPTL00923.250EA
0
EUR
CHMPTL00923.250
MAYBTL00923.1
Methyl-4-methoxy-3-nitrobenzoat
Methyl-4-methoxy-3-nitrobenzoat
Formel:
C₉H₉NO₅ Molecular Weight: 211.17 g/mol |
MDL:
MFCD00024213 CAS-Nummer: 40757-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...