Allylphenylsulfon

Lieferant: CHEMPUR

Synonyme: (Allylsulfonyl)benzol

XBX00149.250 XBX00149.1
CHMPXBX00149.250EA 0 EUR
CHMPXBX00149.250 MAYBXBX00149.1
Allylphenylsulfon
Allylphenylsulfon
Formel: C₉H₁₀O₂S
Molecular Weight: 182,24 g/mol
MDL: MFCD00014739
CAS-Nummer: 16212-05-8
EINECS: 240-338-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR