2-Fluor-p-anisidin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluor-4-methoxyanilin Hydrochlorid, 2-Fluor-4-anisidin Hydrochlorid, 4-Amino-3-fluoranisol Hydrochlorid, 2-Fluor-4-methoxyaniliniumchlorid

AN-1030-5G AN-1030-1G
COBBAN-1030-5GEA 76.1 EUR
COBBAN-1030-5G COBBAN-1030-1G
2-Fluor-p-anisidin Hydrochlorid
2-Fluor-p-anisidin Hydrochlorid
Formel: C₇H₉ClFNO
MDL: MFCD01632190
CAS-Nummer: 874959-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR