4-Brom-3-chlor-6-nitroanisol

Lieferant: COMBI-BLOCKS

AN-1108-5G AN-1108-25G AN-1108-1G
COBBAN-1108-5GEA 303 EUR
COBBAN-1108-5G COBBAN-1108-25G COBBAN-1108-1G
4-Brom-3-chlor-6-nitroanisol
4-Brom-3-chlor-6-nitroanisol
Formel: C₇H₅BrClNO₃
Molecular Weight: 266.48 g/mol
MDL: MFCD09027196
CAS-Nummer: 917562-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR