Methyl-5-methoxy-2-nitrobenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
5-Methoxy-2-nitrobenzoesäuremethylester
AN-1112-25G
AN-1112-5G
AN-1112-1G
COBBAN-1112-25GEA
739
EUR
COBBAN-1112-25G
COBBAN-1112-5G
COBBAN-1112-1G
Methyl-5-methoxy-2-nitrobenzoat
Methyl-5-methoxy-2-nitrobenzoat
Formel:
C₉H₉NO₅ Molecular Weight: 211.17 g/mol |
MDL:
MFCD00662902 CAS-Nummer: 2327-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...