4-Nitrophenylhydrazinhydrochlorid ≥98%
Lieferant: COMBI-BLOCKS
Synonyme:
1-(4-Nitrophenyl)hydrazin Hydrochlorid, 4-Nitrophenylhydraziniumchlorid, 4-Nitrophenylhydrazin Hydrochlorid
AN-1288-25G
AN-1288-100G
COBBAN-1288-25GEA
37.5
EUR
COBBAN-1288-25G
COBBAN-1288-100G
4-Nitrophenylhydrazinhydrochlorid ≥98%
4-Nitrophenylhydrazinhydrochlorid
Formel:
C₆H₇N₃O₂·HCl Molecular Weight: 189,6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040592 CAS-Nummer: 636-99-7 EINECS: 211-273-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...