2-Brom-4-fluor-6-nitroanisol
Lieferant: COMBI-BLOCKS
Synonyme:
1-Bromo-5-fluoro-2-methoxy-3-nitrobenzene
AN-1298-5G
AN-1298-1G
COBBAN-1298-5GEA
303
EUR
COBBAN-1298-5G
COBBAN-1298-1G
2-Brom-4-fluor-6-nitroanisol
2-Brom-4-fluor-6-nitroanisol
Formel:
C₇H₅BrFNO₃ Molecular Weight: 250.02 g/mol |
MDL:
MFCD10699605 CAS-Nummer: 179897-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...