2-Brom-6-chlor-4-nitroanisol

Lieferant: COMBI-BLOCKS

AN-1457-5G AN-1457-1G
COBBAN-1457-5GEA 0 EUR
COBBAN-1457-5G COBBAN-1457-1G
2-Brom-6-chlor-4-nitroanisol
2-Brom-6-chlor-4-nitroanisol
Formel: C₇H₅BrClNO₃
MDL: MFCD15143447
CAS-Nummer: 1215205-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR