4-Bromphenylhydraziniumchlorid
Lieferant: COMBI-BLOCKS
Synonyme:
1-(4-Bromphenyl)hydrazin Hydrochlorid, 4-Bromphenylhydrazinhydrochlorid, 4-Bromphenylhydrazin Hydrochlorid
AN-1544-5G
AN-1544-25G
AN-1544-100G
COBBAN-1544-5GEA
26.2
EUR
COBBAN-1544-5G
COBBAN-1544-25G
COBBAN-1544-100G
4-Bromphenylhydraziniumchlorid
4-Bromphenylhydraziniumchlorid
Formel:
C₆H₈BrClN₂ Molecular Weight: 223,50 g/mol |
MDL:
MFCD00012941 CAS-Nummer: 622-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...