2-Fluor-p-anisidin
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluor-4-anisidin, 4-Amino-3-fluoranisol, 2-Fluor-4-methoxyanilin
AN-2546-1G
AN-2546-5G
COBBAN-2546-1GEA
22
EUR
COBBAN-2546-1G
COBBAN-2546-5G
2-Fluor-p-anisidin
2-Fluor-p-anisidin
Formel:
C₇H₈FNO Molecular Weight: 141,15 g/mol |
MDL:
MFCD00047827 CAS-Nummer: 458-52-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...