2,4-Dibrom-6-methoxyanilin
Lieferant: COMBI-BLOCKS
Synonyme:
4,6-Dibrom-o-anisidin, 2-Amino-3,5-dibromanisol
AN-2549-5G
AN-2549-1G
COBBAN-2549-5GEA
365
EUR
COBBAN-2549-5G
COBBAN-2549-1G
2,4-Dibrom-6-methoxyanilin
2,4-Dibrom-6-methoxyanilin
Formel:
C₇H₇Br₂NO |
MDL:
MFCD11977394 CAS-Nummer: 88149-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...