5-Brom-2-fluor-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Brom-2-fluor-3-pyridinylboronsäure, 5-Brom-2-fluorpyridin-3-boronsäure
BB-2660-5G
BB-2660-25G
BB-2660-1G
COBBBB-2660-5GEA
98.3
EUR
COBBBB-2660-5G
COBBBB-2660-25G
COBBBB-2660-1G
5-Brom-2-fluor-3-pyridinboronsäure
5-Bromo-2-fluoro-3-pyridineboronic acid
Formel:
C₅H₄BBrFNO₂ Molecular Weight: 219,8 g/mol |
MDL:
MFCD04039317 CAS-Nummer: 501435-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...