5-Brom-2-methoxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-methoxypyridin-4-boronsäure, 5-Brom-2-methoxy-4-pyridinylboronsäure

BB-3870-5G BB-3870-1G
COBBBB-3870-5GEA 1180 EUR
COBBBB-3870-5G COBBBB-3870-1G
5-Brom-2-methoxy-4-pyridinboronsäure
5-Bromo-2-methoxy-4-pyridineboronic acid
Formel: C₆H₇BBrNO₃
Molecular Weight: 231.84 g/mol
MDL: MFCD07781223
CAS-Nummer: 957060-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR