6-Brom-4-methyl-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-Brom-4-methylpyridin-3-boronsäure, 6-Brom-4-methyl-3-pyridinylboronsäure

BB-3991-100M BB-3991-250M
COBBBB-3991-100MEA 303 EUR
COBBBB-3991-100M COBBBB-3991-250M
6-Brom-4-methyl-3-pyridinboronsäure
6-Bromo-4-methyl-3-pyridineboronic acid
Formel: C₆H₇BBrNO₂
Molecular Weight: 215.84 g/mol
MDL: MFCD09037489
CAS-Nummer: 1072945-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR