4-Brom-6-chlor-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-6-chlor-3-pyridinylboronsäure, 4-Brom-6-chlorpyridin-3-boronsäure

BB-5077-1G BB-5077-250M
COBBBB-5077-1GEA 1580 EUR
COBBBB-5077-1G COBBBB-5077-250M
4-Brom-6-chlor-3-pyridinboronsäure
4-Bromo-6-chloro-3-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236.26 g/mol
MDL: MFCD09878338
CAS-Nummer: 957062-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR