2-Brom-5-fluor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-5-fluorpyridin-4-boronsäure, 2-Brom-5-fluor-4-pyridinylboronsäure

BB-5320-5G BB-5320-1G
COBBBB-5320-5GEA 181 EUR
COBBBB-5320-5G COBBBB-5320-1G
2-Brom-5-fluor-4-pyridinboronsäure
2-Bromo-5-fluoro-4-pyridineboronic acid
Formel: C₅H₄BBrFNO₂
Molecular Weight: 219,8 g/mol
MDL: MFCD04973353
CAS-Nummer: 1072951-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR