3-Brom-2-fluor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-2-fluorpyridin-4-boronsäure, 3-Brom-2-fluor-4-pyridinylboronsäure

BB-9023-5G BB-9023-1G
COBBBB-9023-5GEA 884 EUR
COBBBB-9023-5G COBBBB-9023-1G
3-Brom-2-fluor-4-pyridinboronsäure
3-Bromo-2-fluoro-4-pyridineboronic acid
Formel: C₅H₄BBrFNO₂
MDL: MFCD11855855
CAS-Nummer: 1150114-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR