(4-Bromphenyl)methylsulfon

Lieferant: COMBI-BLOCKS

BK-4512-100G BK-4512-25G
COBBBK-4512-100GEA 0 EUR
COBBBK-4512-100G COBBBK-4512-25G
(4-Bromphenyl)methylsulfon
(4-Bromphenyl)methylsulfon
Formel: C₇H₇BrO₂S
MDL: MFCD00025065
CAS-Nummer: 3466-32-8
EINECS: 222-421-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR