4-Brom-3-methoxybenzoesäure
Lieferant: COMBI-BLOCKS
CA-4191-1G
CA-4191-5G
COBBCA-4191-1GEA
68.2
EUR
COBBCA-4191-1G
COBBCA-4191-5G
4-Brom-3-methoxybenzoesäure
4-Bromo-3-methoxybenzoic acid
Formel:
C₈H₇BrO₃ |
MDL:
MFCD08436017 CAS-Nummer: 56256-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...