Methyl-2-brom-5-methoxybenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-5-methoxybenzoesäuremethylester

CA-4294-100G CA-4294-5G CA-4294-25G
COBBCA-4294-100GEA 682 EUR
COBBCA-4294-100G COBBCA-4294-5G COBBCA-4294-25G
Methyl-2-brom-5-methoxybenzoat
Methyl-2-brom-5-methoxybenzoat
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
MDL: MFCD00051594
CAS-Nummer: 35450-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR