5-Brom-2-ethoxypyridin

Lieferant: COMBI-BLOCKS

CA-4326-5G CA-4326-25G
COBBCA-4326-5GEA 51.3 EUR
COBBCA-4326-5G COBBCA-4326-25G
5-Brom-2-ethoxypyridin
5-Brom-2-ethoxypyridin
Formel: C₇H₈BrNO
Molecular Weight: 202,05 g/mol
MDL: MFCD00234311
CAS-Nummer: 55849-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR