Methyl-4-methoxy-2-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methoxy-2-nitrobenzoesäuremethylester

CA-4467-1G CA-4467-5G
COBBCA-4467-1GEA 127 EUR
COBBCA-4467-1G COBBCA-4467-5G
Methyl-4-methoxy-2-nitrobenzoat
Methyl-4-methoxy-2-nitrobenzoat
Formel: C₉H₉NO₅
Molecular Weight: 211.17 g/mol
MDL: MFCD06658482
CAS-Nummer: 181871-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR