Methyl-4-chlor-2-methoxybenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
4-Chlor-2-methoxybenzoesäuremethylester
CA-4474-1G
CA-4474-5G
COBBCA-4474-1GEA
101
EUR
COBBCA-4474-1G
COBBCA-4474-5G
Methyl-4-chlor-2-methoxybenzoat
Methyl-4-chlor-2-methoxybenzoat
Formel:
C₉H₉ClO₃ Molecular Weight: 200,62 g/mol |
MDL:
MFCD09800922 CAS-Nummer: 78955-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...