Methyl-3-methoxy-4-nitrobenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
3-Methoxy-4-nitrobenzoesäuremethylester
CA-4640-5G
CA-4640-25G
CA-4640-1G
COBBCA-4640-5GEA
259
EUR
COBBCA-4640-5G
COBBCA-4640-25G
COBBCA-4640-1G
Methyl-3-methoxy-4-nitrobenzoat
Methyl-3-methoxy-4-nitrobenzoat
Formel:
C₉H₉NO₅ Molecular Weight: 211,17 g/mol |
MDL:
MFCD00051967 CAS-Nummer: 5081-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...