Methyl-4-brom-2-methoxybenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-2-methoxybenzoesäuremethylester

CA-4658-5G CA-4658-25G CA-4658-1G
COBBCA-4658-5GEA 88.6 EUR
COBBCA-4658-5G COBBCA-4658-25G COBBCA-4658-1G
Methyl-4-brom-2-methoxybenzoat
Methyl-4-brom-2-methoxybenzoat
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
MDL: MFCD06797867
CAS-Nummer: 139102-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR