3-Brom-5-methoxybenzoesäure

Lieferant: COMBI-BLOCKS

CA-4755-1G CA-4755-5G
COBBCA-4755-1GEA 38.2 EUR
COBBCA-4755-1G COBBCA-4755-5G
3-Brom-5-methoxybenzoesäure
3-Bromo-5-methoxybenzoic acid
Formel: C₈H₇BrO₃
MDL: MFCD00266858
CAS-Nummer: 157893-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR