Methyl-3-brom-4-methoxybenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-4-methoxybenzoesäuremethylester

CA-4789-5G CA-4789-1G
COBBCA-4789-5GEA 98.3 EUR
COBBCA-4789-5G COBBCA-4789-1G
Methyl-3-brom-4-methoxybenzoat
Methyl-3-brom-4-methoxybenzoat
Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
MDL: MFCD00210463
CAS-Nummer: 35450-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR