Methyl-2-brom-6-methoxybenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
2-Brom-6-methoxybenzoesäuremethylester
CA-4963-5G
CA-4963-1G
COBBCA-4963-5GEA
1910
EUR
COBBCA-4963-5G
COBBCA-4963-1G
Methyl-2-brom-6-methoxybenzoat
Methyl-2-brom-6-methoxybenzoat
Formel:
C₉H₉BrO₃ Molecular Weight: 245,07 g/mol |
MDL:
MFCD14697992 CAS-Nummer: 31786-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...