2-Hydroxy-3-(trifluormethyl)pyridin
Lieferant: COMBI-BLOCKS
Synonyme:
3-(Trifluormethyl)pyridin-2-ol
OS-7421-5G
OS-7421-25G
OS-7421-1G
OS-7421-100G
COBBOS-7421-5GEA
26.2
EUR
COBBOS-7421-5G
COBBOS-7421-25G
COBBOS-7421-1G
COBBOS-7421-100G
2-Hydroxy-3-(trifluormethyl)pyridin
2-Hydroxy-3-(trifluoromethyl)pyridine
Formel:
C₆H₄F₃NO Molecular Weight: 163.1 g/mol |
MDL:
MFCD00153194 CAS-Nummer: 22245-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...