2-Fluor-3-methoxybenzoesäure

Lieferant: COMBI-BLOCKS

OT-0033-25G OT-0033-5G OT-0033-1G
COBBOT-0033-25GEA 385 EUR
COBBOT-0033-25G COBBOT-0033-5G COBBOT-0033-1G
2-Fluor-3-methoxybenzoesäure
2-Fluoro-3-methoxybenzoic acid
Formel: C₈H₇FO₃
MDL: MFCD02179621
CAS-Nummer: 137654-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR