Methyl-m-anisat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl-3-methoxybenzoat, 3-Anissäuremethylester, 3-Methoxybenzoesäuremethylester, m-Anissäuremethylester

OT-0753-5G OT-0753-25G
COBBOT-0753-5GEA 37.5 EUR
COBBOT-0753-5G COBBOT-0753-25G
Methyl-m-anisat
Methyl-m-anisat
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00017193
CAS-Nummer: 5368-81-0
EINECS: 226-359-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR