2,4,5-Trifluorthioanisol
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl(2,4,5-trifluorophenyl)sulphane
OT-0854-5G
OT-0854-1G
OT-0854-250M
COBBOT-0854-5GEA
1810
EUR
COBBOT-0854-5G
COBBOT-0854-1G
COBBOT-0854-250M
2,4,5-Trifluorthioanisol
2,4,5-Trifluorthioanisol
Formel:
C₇H₅F₃S Molecular Weight: 178,18 g/mol |
MDL:
MFCD12026061 CAS-Nummer: 54378-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...