2-Brom-5-fluor-4-iodpyridin

Lieferant: COMBI-BLOCKS

PY-1323-1G PY-1323-5G PY-1323-250M
COBBPY-1323-1GEA 497 EUR
COBBPY-1323-1G COBBPY-1323-5G COBBPY-1323-250M
2-Brom-5-fluor-4-iodpyridin
2-Bromo-5-fluoro-4-iodopyridine
Formel: C₅H₂BrFIN
MDL: MFCD15143627
CAS-Nummer: 1061357-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR