2-Hydroxy-5-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-(Trifluormethyl)pyridin-2-ol

PY-7192-25G PY-7192-5G
COBBPY-7192-25GEA 102 EUR
COBBPY-7192-25G COBBPY-7192-5G
2-Hydroxy-5-(trifluormethyl)pyridin
2-Hydroxy-5-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NO
Molecular Weight: 163,1 g/mol
MDL: MFCD00042315
CAS-Nummer: 33252-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR