4-Brom-2-fluorbenzaldehyd

Lieferant: FLUOROCHEM
Achtung

003062-25G 003062-10G 003062-100G
FLUO003062-25GEA 45.9 EUR
FLUO003062-25G FLUO003062-10G FLUO003062-100G
4-Brom-2-fluorbenzaldehyd
4-Bromo-2-fluorobenzaldehyde
Formel: C₇H₄BrFO
Molecular Weight: 203.01 g/mol
MDL: MFCD00143261
CAS-Nummer: 57848-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR