2,2,2-Trifluorethylacrylat

Lieferant: FLUOROCHEM

Synonyme: Acrylsäure(2,2,2-trifluorethyl)ester, Propensäure(2,2,2-trifluorethyl)ester

001250-25G 001250-100G 001250-10G
FLUO001250-25GEA 121 EUR
FLUO001250-25G FLUO001250-100G FLUO001250-10G
2,2,2-Trifluorethylacrylat
2,2,2-Trifluorethylacrylat
Formel: C₅H₅F₃O₂
Molecular Weight: 154,09 g/mol
MDL: MFCD00000444
CAS-Nummer: 407-47-6
EINECS: 206-987-6
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR