(4-Fluorphenyl)methylsulfon

Lieferant: FLUOROCHEM

Synonyme: 1-Fluor-4-(methylsulfonyl)benzol

001765-5G 001765-25G
FLUO001765-5GEA 48.8 EUR
FLUO001765-5G FLUO001765-25G
(4-Fluorphenyl)methylsulfon
(4-Fluorphenyl)methylsulfon
Formel: C₇H₇FO₂S
Molecular Weight: 174.2 g/mol
MDL: MFCD00039753
CAS-Nummer: 455-15-2
EINECS: 207-237-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR