Nitrosiumhexafluorophosphat

Lieferant: FLUOROCHEM

Synonyme: Nitrosylhexafluorophosphat

002854-5G 002854-25G
FLUO002854-5GEA 186 EUR
FLUO002854-5G FLUO002854-25G
Nitrosiumhexafluorophosphat
Nitrosium hexafluorophosphate
Formel: F₆NOP
Molecular Weight: 174,97 g/mol
MDL: MFCD00040324
CAS-Nummer: 16921-91-8
EINECS: 240-980-9
UN: 3260
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR